SCS110 competent cells
Leverancier: STRATAGENE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
200247 200247
STRA200247EA 0 EUR
STRA200247 AGLS200247
SCS110 competent cells
Competente cellen
SCS110 competent cells for generating unmethylated DNA improve quality and yield of miniprep DNA.
  • Transformation efficiency of ≥5×10⁶ transformants/μg DNA
  • Propagated DNA digestible by methylation-sensitive enzymes such as Xba I, Cla I, and EcoR II;Endonuclease-deficient cells (EndA-) improve miniprep quality
  • Blue-white screening for recombinant plasmids
  • Propagated DNA digestible by methylation-sensitive enzymes such as Xba I, Cla I, and EcoR II
SCS110 competent cells for generating unmethylated DNA are also an EndA- variant, which greatly improves theMore Product Information