XL10-Gold ultracompetent cells
Leverancier: STRATAGENE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
200314 200314 200315
STRA200314EA 0 EUR
STRA200314 AGLS200314 AGLS200315
XL10-Gold ultracompetent cells
Competente cellen
XL10-Gold ultracompetent cells transform large plasmids and ligated DNA with highest efficiency.
  • Engineered to transform large plasmids and ligated DNA more efficiently than other cell lines
  • Includes Hte phenotype for 20-fold to 30-fold higher transformation efficiency of ligated DNA and better competent cell performance
  • Chemically competent cells that allow efficient transformation of large DNA molecules, including expression vectors and genomic DNA
  • Ideal for plasmid library construction by decreasing bias against larger plasmids and producing more
More Product Information