AriaMx Real-Time PCR systeem

Leverancier: AGILENT
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

5190-7702 5190-7708 5190-9370 600906 600907 401425 401427 401490 401491 401492 401493 401494 G5380AA G5381AA G8830-67001 G8830-67002 G8830-67003 G8830-67004 G8830-67005 G8830-67006 401428
AGLS5190-7702EA 620 EUR
AGLS5190-7702 AGLS5190-7708 AGLS5190-9370 AGLS600906 AGLS600907 AGLS401425 AGLS401427 AGLS401490 AGLS401491 AGLS401492 AGLS401493 AGLS401494 732-3776 732-3777 732-3778 732-3779 732-3780 732-3781 732-3782 732-3783 AGLS401428
AriaMx Real-Time PCR systeem
PCR-toestellen
Het AriaMx Real-Time PCR-systeem is een volledig geïntegreerd systeem voor kwantitatieve PCR-amplificatie, detectie en gegevensanalyse systeem.

  • Gene expression analysis
  • New genotyping/HRM capability, mRNA quantification
  • NGS quantification: Library preparation, result validation, nucleic acid monitoring and rare allele detection
  • Can use self-installable optical cartridges
  • Operates at a Cq uniformity standard deviation of <0,20 at fast cycling (5 s at 95 °C/10 s at 60 °C)
  • Data acquisition time of <3 s/plate
  • Dynamic range of nine orders of magnitude
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR