Bioanalyzer Parts and Accessories
Leverancier: AGILENT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5065-4401 5065-4413 5065-9951
AGLS5065-4401EA 1260 EUR
AGLS5065-4401 AGLS5065-4413 AGLS5065-9951
Bioanalyzer Parts and Accessories
Elektroforesesystemen Systemen voor capillaire elektroforese
Bioanalyzer spare parts, including electrode cartridges, priming stations, filters, and fuses.
  • Removable electrode cartridges allow labs to buy additional cartridges dedicated for specific applications
  • Replacement test chip kits are available for troubleshooting and running diagnostic tests
  • The chips are reusable and expire after three years
  • Multiple chip priming stations are available and may be useful for core facilities or labs with multiple Bioanalyser users
  • To ensure proper maintenance of the Bioanalyzer system additional gaskets are available in the
More Product Information
Order Now
Specificaties
Environmentally Preferable