Bioanalyzer Protein Analysis
Leverancier: AGILENT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5067-1515 5067-1516 5067-1517 5067-1518
AGLS5067-1515EA 1350 EUR
AGLS5067-1515 AGLS5067-1516 AGLS5067-1517 AGLS5067-1518
Bioanalyzer Protein Analysis
Elektroforesesystemen Systemen voor capillaire elektroforese
Electrophoretic separation of proteins and peptides up to 230 kD.
  • Flexibility to choose the appropriate kit based on the sizing range
  • Minimal sample consumption is required, with only 4 µl of sample needed for an accurate analysis
  • Results are achieved rapidly, with the automated analysis of 10 samples performed in approximately 30 minutes
  • Complete data analysis may be performed in one assay, providing sizing, quantitation, and purity information
  • Provides sensitivity comparable to coomassie blue stains across a large linear dynamic range
  • Meet GMP and GLP
More Product Information