Multiple affinity removal starter reagent kits
Leverancier: AGILENT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5185-5986 5185-5987 5185-5988 5185-5991
AGLS5185-5986EA 797 EUR
AGLS5185-5986 AGLS5185-5987 AGLS5185-5988 AGLS5185-5991
Multiple affinity removal starter reagent kits
Eiwitzuivering Protein Purification Kits
Starter reagent kit for multiple affinity removal LC columns.
Leveringsinformatie: Includes: 2 bottles buffer A (1 L), 1 bottle buffer B (1 L), 2 packs of 25 spin filters, 1 pack of 25 spin concentrator.
Order Now
Specificaties
Environmentally Preferable