Multiple affinity removal spin cartridge reagent kit
Leverancier: AGILENT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5188-5254 5188-5252 5188-5249 5188-5250 5188-5251 5188-5252 5188-5254
HEWL5188-5254EA 0 EUR
HEWL5188-5254 HEWL5188-5252 AGLS5188-5249 AGLS5188-5250 AGLS5188-5251 AGLS5188-5252 AGLS5188-5254
Multiple affinity removal spin cartridge reagent kit
Eiwitzuivering Protein Purification Kits
The affinity removal spin cartridge reagent kit is designed to remove the most interfering high-abundant protein (albumin) from human samples (such as plasma, serum, urine or cerebrospinal fluid).
Leveringsinformatie: Kit includes Buffer A, 1 bottle (1 L) Buffer B, 1 bottle (1 L), Spin filters, 2 packs, Spin concentrators, 5 KDa MWCo (4 ml) 1 pack, Luer-Lok adapter 1 pack, Luer-Lok syringes (5 ml) 1 pack, Microtubes, 6 packs, Caps and plugs 1 pack, PTFE Luer-Lok needles 1 pack.
Order Now
Specificaties
Environmentally Preferable