Master mixes, brilliant HRM ultra-fast loci
Leverancier: AGILENT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5190-7827 5190-7909 600536
AGLS5190-7827EA 207 EUR
AGLS5190-7827 AGLS5190-7909 AGLS600536
Master mixes, brilliant HRM ultra-fast loci
Reagentia voor nucleïnezuren qPCR/RT-qPCR Enzymes and Kits
The Brilliant HRM Ultra-Fast Loci Master Mix for High Resolution Melt (HRM) analysis.
  • MgCl₂ and dNTPs are included at optimised concentrations
  • Master Mix is stable after multiple freeze thaws, reducing wastage and increasing batch-to-batch reliability
  • Start Taq polymerase enzyme requires only three minutes for activation
  • The dye is non-toxic and can be added at saturating concentrations - insuring minimal inhibition while preserving high sensitivity
  • Master Mix can be used with the AriaMx Real-Time PCR System or any HRM-capable thermal cycler
The Master Mix combines a mutant Fast-Start Taq polymerase, optimised concentrations of MgCl₂ and dNTPs, and EvaGreen, release-on-demand dye to enable researchers to mix-and-go faster and with total confidence - even for the most difficult genotypes.
The optimised MgCl₂ and dNTP concentrations reduce assay preparation time, while low surfactant levels minimise bubble trouble during pipetting. In addition, the 12-month storage at ‒20 °C is more than double that of other HRM reagents.
The passive reference dye may be added to fluorescence-based quantitative PCR assays to compensate for non-PCR related variations in fluorescence.
Opgelet: For research use only. Not for use in diagnostic procedures.
Order Now
Specificaties
Environmentally Preferable
Applicatie RT-qPCR dye