Fusion DNA polymerases, Herculase II
Leverancier: AGILENT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
600675 600677 600679
AGLS600675EA 67 EUR
AGLS600675 AGLS600677 AGLS600679
Fusion DNA polymerases, Herculase II
Reagentia voor nucleïnezuren End-point PCR Enzymes and Kits
Herculase II Fusion DNA Polymerases produce superior yields with extension times as short as 15 s/kb, even with genomic DNA targets up to 23 kb.
Herculase II helps overcome PCR challenges with successful amplification of targets of all complexity. Easily amplify low abundance, high GC, and other difficult targets with robust yields.