BAC Kits
Leverancier: AGILENT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
FP-1003-0275 FP-1004-0275
AGLSFP-1003-0275EA 1620 EUR
AGLSFP-1003-0275 AGLSFP-1004-0275
BAC Kits
Elektroforesesystemen Systemen voor capillaire elektroforese
Quality control of BAC digests and large DNA fragments using pulsed-field capillary electrophoresis.
  • Reliable sizing - Accurate and precise sizing of large DNA fragments from 75 bp to 48 kb or 75 bp to 165 kb
  • High resolution separations - Optimised pulsed-field method prevents peak broadening, allowing separation of similarly sized large DNA fragments
  • Flag analysis - ProSize data analysis software allows for automated flagging of DNA fragments decreasing analysis times
  • Low sample volume - Only 2 µl required, conserving sample for downstream analysis
  • Fast
More Product Information