Disposable Tip Pipette Heads

Leverancier: AGILENT
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

G5055G G5056G G5057G
AGLSG5055GEA 49260 EUR
AGLSG5055G AGLSG5056G AGLSG5057G
Disposable Tip Pipette Heads
Pipettips
Agilent pipette heads are designed to be easily mounted or exchanged and to offer the advantage of being more forgiving than metal-tipped, fixed-tip heads during collisions, increasing uptime during the life of your instrument.

  • Designed to work in two liquid volume ranges: small transfer (ST) and large transfer (LT)
  • Typical %CVs for dispensing 2 µl are 2,1% using DMSO
  • Compatible with most common life science laboratory reagents and, with optimisation, CVs better than 5% can be achieved across many liquid types

Disposable-tip pipette heads can aspirateMore Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR