ZAG dsDNA Kits
Leverancier: AGILENT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
ZAG-105-5000 ZAG-110-5000 ZAG-130-5000 ZAG-135-5000
AGLSZAG-105-5000EA 2330 EUR
AGLSZAG-105-5000 AGLSZAG-110-5000 AGLSZAG-130-5000 AGLSZAG-135-5000
ZAG dsDNA Kits
Elektroforesesystemen Systemen voor capillaire elektroforese
High-throughput PCR fragment sizing and microsatellite detection.
  • Broad concentration range - All kits have a concentration range of 0,5 to 50 ng/µl
  • Reliable sizing - Four different kits to choose from with a sizing range varying from 1 to 20000 bp
  • Low sample volume - Only 2 µl of sample is required, minimising precious sample loss to QC steps
  • Quick separations - Run times as short as 30 minutes
  • High resolution - Separate PCR fragments under 300 bp with a 3 bp difference or greater
The ZAG dsDNA kits provide a high-throughput analysis solution forMore Product Information