Cleanroom Coveralls with Integrated Hood
Leverancier: ALSICO HIGHTECH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1036987-XS 1036987-S 1036987-M 1036987-L 1036987-XL 1036987-XXL 1036987-3XL 1037223-XS 1037223-S 1037223-M 1037223-L 1037223-XL 1037223-XXL 1037223-3XL 1036993-XS 1036993-S 1036993-M 1036993-L 1036993-XL 1036993-XXL 1036993-3XL 1037224-XS 1037224-S 1037224-M 1037224-L 1037224-XL 1037224-XXL 1037224-3XL
ALSO1036987-XSEA 93 EUR
ALSO1036987-XS ALSO1036987-S ALSO1036987-M ALSO1036987-L ALSO1036987-XL ALSO1036987-XXL ALSO1036987-3XL ALSO1037223-XS ALSO1037223-S ALSO1037223-M ALSO1037223-L ALSO1037223-XL ALSO1037223-XXL ALSO1037223-3XL ALSO1036993-XS ALSO1036993-S ALSO1036993-M ALSO1036993-L ALSO1036993-XL ALSO1036993-XXL ALSO1036993-3XL ALSO1037224-XS ALSO1037224-S ALSO1037224-M ALSO1037224-L ALSO1037224-XL ALSO1037224-XXL ALSO1037224-3XL
Cleanroom Coveralls with Integrated Hood
Werkkleding
These cleanroom coveralls are made of Alsistrong® (98% polyester and 2% carbon). Suitable for the highest cleanroom classes ISO 4 to 9 or GMP classes A to D.
  • Integrated hood with studs
  • Strap with buckle at the back of the hood
  • Concealed zip front fastening
  • Raglan sleeves with anti-static cuffs
  • Studs for name tag on left chest
  • Adjustable elastic in the back
  • Studs on each side of the leg to attach boots
  • Studs down the legs for adjustment or anti-static cuffs down the legs
  • Marker fabric
  • No pockets
Coveralls are available with studs down the garment legsMore Product Information