MAGMA T-Shirts
Leverancier: ALSICO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1010827-XS 1010827-S 1010827-M 1010827-L 1010827-XL 1010827-2XL 1010827-3XL
ALSW1010827-XSEA 55.4 EUR
ALSW1010827-XS ALSW1010827-S ALSW1010827-M ALSW1010827-L ALSW1010827-XL ALSW1010827-2XL ALSW1010827-3XL
MAGMA T-Shirts
Werkkleding
Flame retardant and anti-static long sleeve t-shirt.
  • Round collar in main fabric
  • Top of back and shoulders doubled in main fabric
  • Ideal as underwear in case of electric arcs
Industrial launderable.
Certificaten: Oeko-Tex® Standard 100; EN 1149-5 2018; EN ISO 11612:2015 (A1, B1, C1, D0, E0, F1).