Guanidinium thiocyanate for biotechnology

Leverancier: Ambion
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Guanidinium sulphocyanide, GTC, Guanidine Thiocyanate, Guanidinium rhodanide

9420 9422
AMBI9420EA 0 EUR
AMBI9420 AMBI9422
Guanidinium thiocyanate for biotechnology
Guanidinium thiocyanate

A powerful protein denaturant used to inactivate endogenous RNases during the isolation of RNA from tissues and bacterial cultures.

Formule: CH₅N₃·CHNS
MW: 118.16 g/mol
Smeltpunt: 118
Dichtheid: 1.29 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00013027
CAS nummer: 593-84-0
EINECS: 209-812-1

Order Now

Specification Test Results

Abs.@280nm (70%, Water) < = 0.8
Abs.@300nm (6M, Water) < 0.1
Abs.@410nm (6M, Water) < 0.1
Copper < 0.0001 %
DNase none detected
Solubility (70%, Water 50° C) pass
Lead < 0.0001 %
Melting Point 118 °C - 121 °C
Protease none detected
Purity > 99 %
RNase none detected


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR