Chemical resistant overalls, AlphaTec® 2000 Standard, Model 111
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
GR20T-00111-03 GR20T-00111-07 GR20T-00111-02 GR20T-00111-04 GR20T-00111-06 GR20T-00111-05 WH20T-00111-08 WH20T-00111-09
ANSEGR20T-00111-03EA 576 EUR
ANSEGR20T-00111-03 ANSEGR20T-00111-07 ANSEGR20T-00111-02 ANSEGR20T-00111-04 ANSEGR20T-00111-06 ANSEGR20T-00111-05 ANSEWH20T-00111-08 ANSEWH20T-00111-09
Chemical resistant overalls, AlphaTec® 2000 Standard, Model 111
Wegwerpkleding
Superior breathable microporous laminate technology, offering Type 5/6 protection.
  • Proven protective barrier to low concentration liquid chemicals, liquid and particulate biological hazards
  • Comfortable moisture vapor permeable fabric helps reduce the risk of heat stress
  • Optimized body fit, improving wearer comfort and safety
  • Anti-static, tested according to EN 1149-5
  • Low linting to help reduce the risk of cross contamination in critical areas
  • Silicone-free, critical in spray painting applications
Coveralls feature a 3-piece hood, 2-way front zipperMore Product Information