AlphaTec® 2000 213 Standard Apron Stitched
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
WH20S-00213-00
ANSEWH20S-00213-00EA 308 EUR
ANSEWH20S-00213-00
AlphaTec® 2000 213 Standard Apron Stitched
Wegwerpkleding
Apron with protective barrier to low concentration liquid chemicals, liquid and particulate biological hazards.
  • Low-linting microporous laminate: Breathable comfort with lower contamination risks
  • Optimised fit: An ideal balance of comfort and safety
Specialised protection: This AlphaTec® apron ensures barrier protection and resists saturation when confronted with low-concentration liquid chemicals, liquid and particulate biological hazards.
Heightened breathability: The use of microporous laminate technology in its fabrication makes for a
More Product Information