Encapsulated chemical protective suits for PAPR systems, AlphaTec® MICROGARD 2500
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
WH25T-00705-05
ANSEWH25T-00705-05EA 0 EUR
ANSEWH25T-00705-05
Encapsulated chemical protective suits for PAPR systems, AlphaTec® MICROGARD 2500
Werkkleding
Encapsulated chemical protective suits for use with PAPR systems.
  • Provide complete protection from liquid and particulate hazards
  • Unique material offering exceptional mechanical strength, as well as liquid and particulate protection
  • Air permeable SMS collar maintains sufficient air in the breathing zone while allowing excess air to flow into the suit body
  • Four exhalation valves allow a full range of movement without risk of excessive pressure
  • Anti-static tested according to EN 1149-5
  • Designed for use with Malina CleanAir fan units (sold separately)