Automated Cell Counter, fluidlab R-300
Leverancier: ANVAJO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
10086 10085 10056 START100 START50
ANVA10086EA 4110 EUR
ANVA10086 ANVA10085 ANVA10056 ANVASTART100 ANVASTART50
Automated Cell Counter, fluidlab R-300
Celtellers Automatische celtellers
The fluidlab R-300 combines an automated cell counter with a full-fledged spectrometer, thus enabling a variety of analyses such as automated cell counting and staining-free viability measurements as well as scatter light, absorbance and time-series measurements. With its intuitive handling and reliable results, the portable device is the all-rounder for your everyday laboratory work.
  • Label-free viability measurements via holographic microscopy
  • Large FoV (5,3 mm²) for high statistical significance
  • Small sample volume needed (10 to 20 µl)
  • Measurement time &
More Product Information