5-Bromo-4-chloro-3-indolyl phenyl phosphonate, p-toluidine salt
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
Waarschuwing
BI5949-100MG BI5949-250MG
APOSBI5949-100MGEA 217 EUR
APOSBI5949-100MG APOSBI5949-250MG
5-Bromo-4-chloro-3-indolyl phenyl phosphonate, p-toluidine salt
5-Bromo-4-chloro-3-indolyl phenyl phosphonate, p-toluidine salt
Chromogenic enzyme substrate for the detection ofphosphonate monoesterase, a conditional lethal gene (pehA) used in the study of plant genetics.