Adenosine 99%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

BIA2545-100G BIA2545-10G BIA2545-25G
APOSBIA2545-100GEA 87.6 EUR
APOSBIA2545-100G APOSBIA2545-10G APOSBIA2545-25G
Adenosine 99%
Adenosine

Endogenous neurotransmitter.

Formule: C₁₀H₁₃N₅O₄
Smeltpunt: 234…236 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00005752
CAS nummer: 58-61-7
EINECS: 200-389-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR