6-Azauracil

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 1,2,4-Triazine-3,5(2H,4H)-dione

BIA4024-100G BIA4024-25G BIA4024-500G
APOSBIA4024-100GEA 110 EUR
APOSBIA4024-100G APOSBIA4024-25G APOSBIA4024-500G
6-Azauracil
6-Azauracil
Formule: C₃H₃N₃O₂
MW: 113.08 g/mol
Smeltpunt: 274…275 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006456
CAS nummer: 461-89-2
EINECS: 207-318-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR