L-Phenylalanine 7-amido-4-methylcoumarin trifluoroacetate salt

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: L-Phenylalanine-AMC.TFA

BIB1443-250MG
APOSBIB1443-250MGEA 87.6 EUR
APOSBIB1443-250MG
L-Phenylalanine 7-amido-4-methylcoumarin trifluoroacetate salt
L-Phenylalanine 7-amido-4-methylcoumarin trifluoroacetate salt
Formule: C₁₉H₁₈N₂O₃·CF₃CO₂H
Smeltpunt: 202…206 °C
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00167024
CAS nummer: 108321-84-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR