L-Proline 7-amido-4-methylcoumarin hydrobromide salt

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

BIB1444-100MG BIB1444-250MG BIB1444-50MG
APOSBIB1444-100MGEA 87.6 EUR
APOSBIB1444-100MG APOSBIB1444-250MG APOSBIB1444-50MG
L-Proline 7-amido-4-methylcoumarin hydrobromide salt
L-Proline 7-amido-4-methylcoumarin hydrobromide salt
Formule: C₁₅H₁₆N₂O₃·BrH
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00213795
CAS nummer: 115388-93-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR