γ-(N-Morpholino)propanesulphonic acid (MOPS) 99,5, ultrapure

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 3-Morpholinopropane-1-sulphonic acid, 3-(N-Morpholino)propanesulphonic acid

BIB2003-1KG BIB2003-250G BIB2003-500G
APOSBIB2003-1KGEA 220 EUR
APOSBIB2003-1KG APOSBIB2003-250G APOSBIB2003-500G
γ-(N-Morpholino)propanesulphonic acid (MOPS) 99,5, ultrapure
γ-(N-Morpholino)propanesulphonic acid (MOPS)

Biological buffer - useful range 6.5-7.9


  • Heavy metals:<5 ppm
  • pKa @ 20°C: 7.20 + 0.2

Formule: C₇H₁₅NO₄S
MW: 209.27 g/mol
Smeltpunt: >277 °C
Vlampunt: 116 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00006183
CAS nummer: 1132-61-2
EINECS: 214-478-5
Merck Index: 13,06288

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR