Blasticidin S hydrochloride

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

BIB4432-50MG
APOSBIB4432-50MGEA 184 EUR
APOSBIB4432-50MG
Blasticidin S hydrochloride
Blasticidin S hydrochloride

Nucleoside antibiotic isolated from Streptomyces griseochromogenes.

Formule: C₁₇H₂₇ClN₈O₅
Smeltpunt: 218…225 °C
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD02091640
CAS nummer: 3513-03-9
UN: 2811
ADR: 6.1,II

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR