2-Keto-D-gluconic acid hemicalcium salt monohydrate

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BIB6086-10G BIB6086-1G BIB6086-5G
APOSBIB6086-10GEA 600 EUR
APOSBIB6086-10G APOSBIB6086-1G APOSBIB6086-5G
2-Keto-D-gluconic acid hemicalcium salt monohydrate
2-Keto-D-gluconic acid hemicalcium salt monohydrate
Formule: C₆H₉O₇·½Ca·H₂O
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00149104
CAS nummer: 3470-37-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR