L(-)-Malic acid disodium salt

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

BIB6095-100G BIB6095-25G
APOSBIB6095-100GEA 176 EUR
APOSBIB6095-100G APOSBIB6095-25G
L(-)-Malic acid disodium salt
L(-)-Malic acid disodium salt
Formule: C₄H₄Na₂O₅
Bewaar temperatuur: Kamertemperatuur
CAS nummer: 138-09-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR