4-Methylumbelliferyl phosphate disodium salt (4-MUP-Na2) trihydrate

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Disodium 4-methylumbelliferyl phosphate trihydrate, 4-MUP trihydrate

BIB6118-1G BIB6118-500MG BIB6118-2.5G
APOSBIB6118-1GEA 125 EUR
APOSBIB6118-1G APOSBIB6118-500MG APOSBIB6118-2.5G
4-Methylumbelliferyl phosphate disodium salt (4-MUP-Na2) trihydrate
4-Methylumbelliferyl phosphate disodium salt (4-MUP-Na2) trihydrate
Formule: C₁₀H₇Na₂O₆P·3H₂O
MW: 354.16 g/mol
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00037614
CAS nummer: 22919-26-2
EINECS: 245-325-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR