Miltefosine

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

BIB6120-1G BIB6120-2.5G BIB6120-250MG
APOSBIB6120-1GEA 139 EUR
APOSBIB6120-1G APOSBIB6120-2.5G APOSBIB6120-250MG
Miltefosine
Miltefosine
Formule: C₂₁H₄₆NO₄P
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00133396
CAS nummer: 58066-85-6
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR