Tetradecyltrimethylammonium bromide BP

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: MiTMAB, (1-Tetradecyl)trimethylammonium bromide, N,N,N-Trimethyltetradecan-1-aminium bromide, Tetradonium bromide, Tetradecyl trimethyl ammonium Bromide, Myristyltrimethylammonium bromide, Dynamin Inhibitor II, TTAB

BIC0114-100G BIC0114-500G
APOSBIC0114-100GEA 49.7 EUR
APOSBIC0114-100G APOSBIC0114-500G
Tetradecyltrimethylammonium bromide BP
Tetradecyltrimethylammonium bromide
Formule: C₁₇H₃₈BrN
MW: 336.394 g/mol
Smeltpunt: 245…250 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011770
CAS nummer: 1119-97-7
UN: 3263
ADR: 8 II
Merck Index: 12,06419

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR