(3-((3-Cholamidopropyl)dimethylammonio)-2-hydroxy-1-propanesulphonate) ≥99.0%, Ultrapure

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

BIC0121-1G BIC0121-5G
APOSBIC0121-1GEA 43.8 EUR
APOSBIC0121-1G APOSBIC0121-5G
(3-((3-Cholamidopropyl)dimethylammonio)-2-hydroxy-1-propanesulphonate) ≥99.0%, Ultrapure
(3-((3-Cholamidopropyl)dimethylammonio)-2-hydroxy-1-propanesulphonate)

A non denaturing zwitterionic detergent similar to CHAPS(BIMB1085) with increased solubility.

Formule: C₃₂H₅₈N₂O₈S
Smeltpunt: 184…186 °C
Bewaar temperatuur: Kamertemperatuur
CAS nummer: 82473-24-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR