EMCH.HCl (E-Maleimidocaproic acid hydrazine hydrochloride)

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

BICL224-250MG BICL224-1G
APOSBICL224-250MGEA 26.3 EUR
APOSBICL224-250MG APOSBICL224-1G
EMCH.HCl (E-Maleimidocaproic acid hydrazine hydrochloride)
EMCH.HCl (E-Maleimidocaproic acid hydrazine hydrochloride)

Sulphhydryl reactive -Carbohydrate reactive

Formule: C₁₀H₁₅N₃O₃·ClH
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD07363434
CAS nummer: 175290-73-0

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR