3,4-Dideoxyglucosone-3-ene ≥98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 3,4-DGE

BICL4066-1MG BICL4066-5MG BICL4066-2MG
APOSBICL4066-1MGEA 520 EUR
APOSBICL4066-1MG APOSBICL4066-5MG APOSBICL4066-2MG
3,4-Dideoxyglucosone-3-ene ≥98%
3,4-Dideoxyglucosone-3-ene
Formule: C₆H₈O₄
MW: 144.125 g/mol
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD02683312
CAS nummer: 252006-38-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR