Gallic acid 3-O-β-D-glucopyranoside ≥95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

BICL4337-10MG BICL4337-25MG BICL4337-5MG
APOSBICL4337-10MGEA 634 EUR
APOSBICL4337-10MG APOSBICL4337-25MG APOSBICL4337-5MG
Gallic acid 3-O-β-D-glucopyranoside ≥95%
Gallic acid 3-O-β-D-glucopyranoside
Formule: C₁₃H₁₆O₁₀
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD28397798
CAS nummer: 91984-84-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR