4-Methoxyphenyl 2,3,6,2',3',4',6'-hepta-O-acetyl-β-D-maltopyranoside ≥98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BICL4357-1G BICL4357-5G BICL4357-2G
APOSBICL4357-1GEA 380 EUR
APOSBICL4357-1G APOSBICL4357-5G APOSBICL4357-2G
4-Methoxyphenyl 2,3,6,2',3',4',6'-hepta-O-acetyl-β-D-maltopyranoside ≥98%
4-Methoxyphenyl 2,3,6,2',3',4',6'-hepta-O-acetyl-β-D-maltopyranoside
Formule: C₃₃H₄₂O₁₉
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD28397804
CAS nummer: 848863-34-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR