Methyl 2,3,6,2',3',4',6'-hepta-O-acetyl-β-D-maltopyranoside ≥98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BICL4361-1G BICL4361-2G BICL4361-500MG
APOSBICL4361-1GEA 774 EUR
APOSBICL4361-1G APOSBICL4361-2G APOSBICL4361-500MG
Methyl 2,3,6,2',3',4',6'-hepta-O-acetyl-β-D-maltopyranoside ≥98%
Methyl 2,3,6,2',3',4',6'-hepta-O-acetyl-β-D-maltopyranoside
Formule: C₂₇H₃₈O₁₈
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD28397808
CAS nummer: 13223-83-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR