Decyl-β-D-glucopyranoside

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

BID6001-1G BID6001-5G BID6001-10G
APOSBID6001-1GEA 96.4 EUR
APOSBID6001-1G APOSBID6001-5G APOSBID6001-10G
Decyl-β-D-glucopyranoside
Decyl-β-D-glucopyranoside
Formule: C₁₆H₃₂O₆
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00063297
CAS nummer: 58846-77-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR