n-Decyl-β-D-maltoside

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

BID6002-1G BID6002-5G BID6002-10G
APOSBID6002-1GEA 103 EUR
APOSBID6002-1G APOSBID6002-5G APOSBID6002-10G
n-Decyl-β-D-maltoside
n-Decyl-β-D-maltoside
Formule: C₂₂H₄₂O₁₁
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00061624
CAS nummer: 82494-09-5

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR