5-Fluorouracil

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Fluorouracil, 5-Fluoropyrimidine-2,4(1H,3H)-dione, 5-FU

BIF0123-25G BIF0123-50G BIF0123-5G
APOSBIF0123-25GEA 70.1 EUR
APOSBIF0123-25G APOSBIF0123-50G APOSBIF0123-5G
5-Fluorouracil
5-Fluorouracil

This pyrimidine analogue, is an antineoplastic agent.

Formule: C₄H₃FN₂O₂
MW: 130.08 g/mol
Kookpunt: 190…200 °C/0.1mmHg
Smeltpunt: 282…286 °C (ontbinding)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00006018
CAS nummer: 51-21-8
EINECS: 200-085-6
UN: 2811
ADR: 6.1 III
Merck Index: 13,04208

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR