Fast green FCF

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BIF1401-10G BIF1401-25G
APOSBIF1401-10GEA 87.6 EUR
APOSBIF1401-10G APOSBIF1401-25G
Fast green FCF
Fast green FCF

C.I. 42053

Formule: C₃₇H₃₄N₂Na₂O₁₀S₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00013053
CAS nummer: 2353-45-9

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR