Riboflavine-5'-phosphate sodium salt USP

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

BIF4012-100G BIF4012-25G BIF4012-1KG
APOSBIF4012-100GEA 512 EUR
APOSBIF4012-100G APOSBIF4012-25G APOSBIF4012-1KG
Riboflavine-5'-phosphate sodium salt USP
Riboflavine-5'-phosphate sodium salt
Formule: C₁₇H₂₀N₄NaO₉P
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00065362
CAS nummer: 130-40-5
EINECS: 204-988-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR