Ruxolitinib free base ≥99%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

BIFK0027-10MG BIFK0027-25MG BIFK0027-5MG
APOSBIFK0027-10MGEA 87.6 EUR
APOSBIFK0027-10MG APOSBIFK0027-25MG APOSBIFK0027-5MG
Ruxolitinib free base ≥99%
Ruxolitinib
Formule: C₁₇H₁₈N₆
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD12031592
CAS nummer: 941678-49-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR