Hydroxyurea 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

BIH0153-10G BIH0153-25G BIH0153-5G
APOSBIH0153-10GEA 43.8 EUR
APOSBIH0153-10G APOSBIH0153-25G APOSBIH0153-5G
Hydroxyurea 98%
Hydroxyurea

Anti-neoplastic. Inactivates ribonucleoside reductase byformation of free radical nitroxide. This blocks synthesis of deoxynucleotides which inhibits DNA synthesis.


  • Protein

Formule: CH₄N₂O₂
Smeltpunt: 135 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00007943
CAS nummer: 127-07-1
EINECS: 204-821-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR