3-Iodo-L-tyrosine 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

BII0193-1G BII0193-5G BII0193-25G
APOSBII0193-1GEA 61.4 EUR
APOSBII0193-1G APOSBII0193-5G APOSBII0193-25G
3-Iodo-L-tyrosine 98%
3-Iodo-L-tyrosine

Inhibitor of tyrosin hydroxylase.

Formule: C₉H₁₀INO₃
Smeltpunt: 196…198 °C
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00002608
CAS nummer: 70-78-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR