Candida antarctica Lipase A
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BIIM1000-100G BIIM1000-10G BIIM1000-25G
APOSBIIM1000-100GEA 530 EUR
APOSBIIM1000-100G APOSBIIM1000-10G APOSBIIM1000-25G
Candida antarctica Lipase A
Enzymen
Covalent on IB-150A. Activity: 2000 TBU/g
  • immobilized on Immobead 150