Jasmonic acid (mixture of isomers)

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

BIJ0379-100MG BIJ0379-250MG
APOSBIJ0379-100MGEA 40.9 EUR
APOSBIJ0379-100MG APOSBIJ0379-250MG
Jasmonic acid (mixture of isomers)
Jasmonic acid (mixture of isomers)

  • Plant tissue culture tested

Formule: C₁₂H₁₈O₃
Dichtheid: 1,07 g/cm³ (25 °C)
Bewaar temperatuur: Koelkast
CAS nummer: 221682-41-3

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR