α-Ketoglutaric acid monosodium salt

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

BIK8011-5G BIK8011-25G BIK8011-50G
APOSBIK8011-5GEA 46.8 EUR
APOSBIK8011-5G APOSBIK8011-25G APOSBIK8011-50G
α-Ketoglutaric acid monosodium salt
α-Ketoglutaric acid monosodium salt
Formule: C₅H₅NaO₅
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00064196
CAS nummer: 22202-68-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR