5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na) ≥98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

BIMB1018-1G BIMB1018-250MG BIMB1018-5G
APOSBIMB1018-1GEA 83.3 EUR
APOSBIMB1018-1G APOSBIMB1018-250MG APOSBIMB1018-5G
5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na) ≥98%
5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na)

Substrate for Alkaline phosphatase.


  • Soluble in water

Formule: C₈H₄BrClNNa₂O₄P
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00036757
CAS nummer: 102185-33-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR