Nitro blue tetrazolium chloride (NBT)

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 5,5'-Diphenyl-3,3'-bis(4-nitrophenyl)-2,2'-(3,3'-dimethoxybiphenyl-4,4'-ylene)ditetrazolium dichloride

BIMB1019-1G BIMB1019-250MG BIMB1019-5G
APOSBIMB1019-1GEA 52.6 EUR
APOSBIMB1019-1G APOSBIMB1019-250MG APOSBIMB1019-5G
Nitro blue tetrazolium chloride (NBT)
Nitro blue tetrazolium chloride (NBT)

NBT is used in conjunction with BCIP (BIMB1004) for colourimetric detection of alkaline phophatase activity in blotting, immuno-histochemical and cytochemistry techniques.

Formule: C₄₀H₃₀Cl₂N₁₀O₆
MW: 817.65 g/mol
Smeltpunt: 189 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00012159
CAS nummer: 298-83-9
EINECS: 206-067-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR